Klon-Zylinder
Lieferant: HILGENBERG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1980003 1740174 1980004
710-0203EA 369 EUR InStock
710-0203 710-0202 HILG1980004
Klon-Zylinder
Zylinder Cloning-Zylinder
Borosilikatglas 3.3
  • Hochwertiges Borosilikatglasringe
  • Thermisch Sterilisiert
  • Wanddicke: 1 mm
Cloning cylinders or rings with ground edges are used to separate the individual cells or cell groups in the nutrient soultion by placing the ring over it.The ring requires little grease to provide an optimum seal between the ring and the petri dish.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge