VWR®, Handstückzähler
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
710-0935EA 39 EUR InStock
710-0935
VWR®, Handstückzähler
Zellzähler und Durchflusszytometer Manuelle Zellzähler
Metal construction is designed for reliability, long life and hard use. Unique engineering provides a smooth tactile feel. Numbers instantly and automatically return to zero with a twist of the reset knob.
  • Compact unit for carrying in the pocket
  • Ease in viewing, large white numerals on a black background
Hundreds of laboratory applications provide opportunities to count blood samples, take surveys, inventory items, total drops of liquid, receive lab samples, and tally biomedical events. A protective window seals out dust, dirt and humidity. The rugged, heavy-duty, chrome-finished case has a hinged finger loop for a secure grip. Finger pressure on the button produces an audible click that signals the count. This feature ensures a registered count without looking at the counter.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge