VWR® Surface Sampling Sponge Kits
Lieferant: VWR Chemicals
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
710-1020EA 769 EUR InStock
710-1020 300-0229 300-0230 300-0231 300-0232 300-0240
VWR® Surface Sampling Sponge Kits
Tupfer Tupfer für die Probenahme
Surface sampling sponges in two forms: Dry sponge sterile sampling bag with write-on strip, containing a dry, biocide-free cellulose sponge, or wet sponge sterile sampling bag, with a foil-sealed, pre-moistened cellulose sponge, and 10 ml of sterile neutralising buffer.
  • Sterilised using gamma irradiation and leakproof transport bags, for the rapid cooling of samples in refrigerators
  • Puncture-proof safety bags eliminating sharp points that can cause puncture and/or abrasions
  • Larger surface in comparison with normal sampling swabs
This surface sampling sponge kit is designed to collect and transport samples, to detect the presence of microbiological contaminations such as Listeria, Salmonella, E. coli, and other food-borne pathogens on almost any surface. These kits are widely used in the food, medical, public health and cosmetic industries.
Zertifizierungen: HACCP, USDA, ACIA and CFIA requirements.
Achtung! Bitte beachten: Gloves should be worn during all sampling procedures.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge