SRK® Surface Control Swabs
Lieferant: COPAN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
906C.CR 906C 9U002S 9U017S 9U011S01 9U012S
710-1976EA 636 EUR InStock
710-1976 710-1975 710-1977 710-2232 710-2231 COPA9U012S
SRK® Surface Control Swabs
Tupfer Tupfer für die Probenahme
Surface control swabs and sponges designed for the hygiene monitoring of surfaces.
  • High recovery for bioburden control
  • Enables validation of sanitization process
Collection and transport systems for microbiological applications in food, hospitals and other sectors such as cosmetics and industries.
Verpackung: VHP resistant triple-protection. Individually wrapped.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge