Lumitester smart, Kikkoman
Lieferant: HYSERVE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1702671 1702672 1902653
710-2244EA 375 EUR InStock
710-2244 710-2245 710-2243
Lumitester smart, Kikkoman
Überwachungssysteme Hygiene-Überwachungssysteme
Luminometer for hygiene validation of surfaces in 10 seconds.
The lumitester Smart, in combination with the LuciPac A3 reagent tests, detects all organic residues and microorganisms on surfaces as well as in liquids and determines the degree of cleanliness at the point of sampling. The system thus enables hygiene monitoring of surfaces within seconds, directly following cleaning and disinfection measures.
The Lumitester Smart is a mobile device that allows it to be used even in places that are difficult to access, making it particularly user-friendly. Via Bluetooth, the device can be connected to smartphones, tablets, and laptops, where comprehensive evaluation of the measurement data is possible. By storing the data in the cloud, the results from multiple users and devices can be combined and evaluated regardless of location.
Lieferumfang: It includes:1 lumitester smart,1 wrist strap, 1 USB cable, 2 batteries (AA, Mignon), 3 cleaning brushes and a quick start guide.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge