Enterobacteriaceae count plates, Petrifilm™
Lieferant: 3M FOOD SAFETY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
6420 6421
710-3402EA 102 EUR InStock
710-3402 710-3403
Enterobacteriaceae count plates, Petrifilm™
Medien für die Mikrobiologie Fertigmedien für die Mikrobiologie
3M™ Petrifilm™ Enterobacteriaceae count plates provide a cost-effective, convenient and reliable method for testing equipment, raw materials, food products and manufacturing environmental samples and bottled water testing.
  • Results in 24 hours
  • Plates are sample-ready, eliminating all the time-consuming steps of preparing media/agar dishes
  • Can be used with the 3M™ Petrifilm™ Plate Reader for consistent, fast, automated reading and recording of results
  • This plate has been awarded Official Methods of Analysis (OMA) #200301 for selected foods and NF VALIDATION certificate: 3M 01/6 to 09/97 for All human food products, Animal feed and Industrial environmental samples by AFNOR Certification
  • Food processors and Contract labs worldwide trust 3M™ Petrifilm™ plates to provide accurate, efficient, consistent results
3M™ Petrifilm™ enterobacteriaceae count plates enable you to quickly determine potential sources of contamination. This plate enumerates all coliforms as well as potential pathogens such as Salmonella, Shigella and Yersini. The 3M™ Petrifilm™ Enterobacteriaceae count plate can also be read using the 3M™ Petrifilm™ plate reader which provides consistent, fast, automated reading and recording of results of 3M™ Petrifilm™ plates (aerobic count, rapid aerobic count, coliform count, E. coli/coliform count, enterobacteriaceae count and select E. coli count ) in 4 seconds.
In today's environment of heightened focus on food safety and increasingly stringent quality requirements, labs are under pressure to provide tests that are fast, reliable and consistently accurate. 3M™ Petrifilm™ plates have demonstrated consistent performance that has been cited globally with more than 200 certificates, recognitions, validations, and peer review publications. For more than 30 years, food safety professionals around the world have put their trust in 3M™ Petrifilm™ plates. That's because 3M™ Petrifilm™ plates have streamlined, standardised and simplified the process of microbial quantitative indicator testing, improving productivity and helping ensure the highest levels of product quality.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge