71125.14
71125.22
71125.14EA
35.9
EUR
InStock
71125.14
71125.22
Butylzinntrichlorid ≥96%
Butylzinntrichlorid
Purity: 96%
CAS No.: 1118-46-3
Molecular Formula: CH3(CH2)3SnCl3
Formula Weight: 282.17
Flash Point: 81°(178°F)
Physical Form: Liquid
Density: 1.693
Refractive Index: 1.5244
Sensitivity: Moisture Sensitive
Odour: Acrid
Melting Point: -63°
Boiling Point: 98°/10mm
UN No.: 3265
MDL No.: MFCD00000515