71193.14
71193.22
71193.36
71193.14EA
27.3
EUR
InStock
71193.14
71193.22
71193.36
Dioctylzinnoxid
Dioctylzinnoxid
CAS No.: 870-08-6
Molecular Formula: (C8H17)2SnO
Formula Weight: 361.14
Physical Form: Powder
Appearance: White
Melting Point: 245-248° dec.
MDL No.: MFCD00013839