D-Desthiobiotin
Lieferant: Novagen (Part of Merck)
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
71610-3
71610-3EA 377 EUR InStock
71610-3
D-Desthiobiotin
Protein-Aufbereitungs Protein Purification Resins
D-Desthiobiotin provides a gentle elution of StrepTag® II proteins from the biotin-binding site of StrepTactin® resins. D-Desthiobiotin is offered as a lyophilised powder for preparation of elution buffer.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge