Ausgießformen zum Einmalgebrauch
Lieferant: Simport Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
M475-1 M475-3 M475-10 M475-4 M475-5 M475-2
720-0820EA 502 EUR InStock
720-0820 720-0822 720-0208 720-0823 720-0824 720-0821
Ausgießformen zum Einmalgebrauch
Formen Histologie-Formen
These disposable PVC base moulds offer ease and convenience of operation. They can be discarded after use, but are also strong enough to be reused.
  • Excellent thermal exchange
  • Smooth interior
  • Rounded corners
They are available in the same variety of sizes as metal moulds and can be used with the same styles or types of cassettes and embedding rings.
Lieferumfang: Each case contains two dispenser boxes of 500 base moulds.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge