Carbolic auramine
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
361430-1000
720-0345EA 69.3 EUR InStock
720-0345
Carbolic auramine
Carbolic auramine
RAL Auramine staining kit allows fluorescence detection of Acid Fast Bacilli (AFB). The A.F.B appear fluorescent-yellow on the blue background preparation.
Used in Auramine staining kit Ref 359500
IVDLogo
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge