Hemaperls kit for haematology
Lieferant: RAL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
362800-0000
720-1184EA 108.65 EUR InStock
720-1184
Hemaperls kit for haematology
Hemaperls kit for haematology
Perls Staining for the Detection of Haemosiderin (Ferric salts - Prussian Blue)
In an acid environment, ferric ions react with potassium ferrocyanide what leads to a precipitate called Ferric Ferrocyanide (Prussian blue). It points out the presence of a pathological pigment: Haemosiderin.
IVDLogo
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge