SupaMega tissue processing cassette
Lieferant: CELLPATH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
EAG-0102-02A
720-1897EA 183 EUR InStock
720-1897
SupaMega tissue processing cassette
Einbettkassetten Gewebeeinbettungskassetten
CellPath’s SupaMega Cassette is ideal for embedding and processing large tissue samples. The SupaMega Cassette is widely used in research and toxicology labratories. Dimension: 65×50×15 mm
  • Ideal for prostate, brain, breast tissue and eyes
  • Extremely useful for research and toxicology laboratories
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge