Automated tissue processor, MTM
Lieferant: Slee
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
11000500
720-1974EA 47040 EUR InStock
720-1974
Automated tissue processor, MTM
Gewebeprozessoren
The MTM II fast tissue processor offers a cost-efficient and user-friendly alternative for fast biopsies processing and overnight batch tissue processing needs of every histology laboratory. The instrument can either be used for fully automated fast biopsy processing or high volume overnight batches of up to 300 samples.
  • Safe and homogeneous tissue processing with preheated reagents (EHE)
  • Fully automatic reagent management system
  • Automatic cleaning cycles
  • Intuitive control via 15'' touchscreen display
  • Charcoal filter supported ventilation
  • Remote alarm system
  • Delayed start function: up to 14 days, 23 hours and 59 minutes
The innovative and patented enhanced heat exchanger (EHE) allows fastest non-microwave tissue processing today available. Reagents are homogeneously and gently heated thereby assuring maximum sample safety.
The system can be operated with traditional reagents or with most of the xylene and ethyl alcohol substitutes today available on the market. An user friendly reagent management system and an optimized cleaning cycle routine are the basis for higher cost efficiency and higher turn-over. Process safety is guaranteed with an intelligent remote alarm system that communicates via mobile or internet.
Reagent filling and draining is either manual or automatic.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge