VWR® Q Path® Safety Pad, Absorbierende Unterlage, Formalinneutralisator-Pads
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
720-2251EA 198 EUR InStock
720-2251 720-2252 720-2253
VWR® Q Path® Safety Pad, Absorbierende Unterlage, Formalinneutralisator-Pads
Biopsieschwämmchen
Absorbs the odours (neutralises formalin). Ensures good contrast for biopsies.
  • Different sizes available
  • Very useful for the bottom of transport boxes and small formalin spills
Protects knives and cutting boards against premature wear.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge