Venenverweilkanülen, Vasofix® Braunüle®
Lieferant: B. Braun
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
4268210B 4268130B 4268091B 4268113B
720-2549EA 108 EUR InStock
720-2549 720-2548 720-2546 720-2547
Venenverweilkanülen, Vasofix® Braunüle®
Katheter
IV catheter material available in PU and FEP for low-pain and gentle insertion. Medication can be quickly administered without having to repuncture the area.
  • Four fully integrated x-ray contrast lines
  • Hydrophobic blood stoppers, removable lock cones
  • Colour-coded sizes
  • Free of latex and PVC
  • Excellent puncture properties with atraumatic tip and triple facet bevel geometry
Same product properties as the Vasofix® Safety (with FEP catheter) excluding the integrated, self-activating safety device, for preventing needlestick injuries and the associated risk of infection.
Zertifizierungen: ISO 10555-1/5.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge