This model has a special hinge that holds the base and the cover together. The hinge design allows the cassette to be opened and closed as often as required. It is a front-opening cassette with a 45° smooth writing surface.
- Unique lateral vents, fluids can access tissue from all four sides
- Basket style for small tissue management
- Special hinged cover design with front opening
- Smooth 45° angle writing surface
- Available in 11 colours
Molded from high-density polymer, keeps specimens safely submerged in liquid.
Available for tissue, biopsy and micro-biopsy. This cassette model is supplied in bulk, in taped stacks for use with Leica and Sakura Ink Jet printers and in plastic sleeves suitable for Thermo Fisher printers.
Zertifizierungen: IVDR, CE.