DNA kit, BacMagic™-2
Lieferant: Novagen (Part of Merck)
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
72157-3
72157-3EA 809 EUR InStock
72157-3
DNA kit, BacMagic™-2
Transfektionsreagenzien
The BacMagic™-2 DNA Kit provides a positive selection for creating baculoviruses recombinants in preparation for generating recombinant protein expression in insect cells. The kit improves on the traditional method for generating recombinant baculoviruses by eliminating the tedious, time-consuming steps of plaque purification.
BacMagic™-2 DNA is derived from an AcNPV genome, contains an ORF 1629 deletion which prevents nonrecombinant virus from replicating in insect cells, and contains a deletion of the non essential chitinase (chiA) gene, to improve production of secreted and membrane-targeted proteins. BacMagic™-2 DNA have an additional deletion of the v-cath gene, a cathespsin-like cysteine protease, resulting in significantly improved quality and yield for most target proteins.
Storage temperature: +2 to +8°C.
Lieferumfang: The BacMagic™-2 DNA Kit contains the BacMagic™-2 DNA, Insect GeneJuice® Transfection Reagent, and a positive control plasmid to monitor co-transfection efficiency.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge