Transfection kit, BacMagic™-2
Lieferant: Novagen (Part of Merck)
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
72158-3
72158-3EA 1100 EUR InStock
72158-3
Transfection kit, BacMagic™-2
Transfektionsreagenzien
The Novagen® BacMagic™-2 transfection kit provides a positive selection for creating baculovirus recombinants in preparation for recombinant protein expression in insect cells. The kit improves on the traditional method for generating recombinant baculoviruses by eliminating the tedious, time-consuming steps of plaque purification.
The Novagen® BacMagic™-2 transfection kit contains BacMagic™-2 DNA, Insect GeneJuice® transfection reagent, and a positive control plasmid to monitor co-transfection efficiency. In addition, it contains Sf9 cells and BacVector® medium, for optimal transfection performance.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge