Colour reference solution acc. to APHA, analytical standard, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
72599-1SET-F 77147-1SET-F
72599-1SET-FEA 470 EUR InStock
72599-1SET-F 77147-1SET-F
Colour reference solution acc. to APHA, analytical standard, Supelco®
Standards Farbreferenzen und Spektroskopie-Standards
This is a set of the following colour reference solutions according to APAH: Option of 2 ml or 10 ml ampoule kits.
Colour Reference Solution 5
Colour Reference Solution 10
Colour Reference Solution 15
Colour Reference Solution 20
Colour Reference Solution 25
Colour Reference Solution 30
Colour Reference Solution 35
Colour Reference Solution 40
Colour Reference Solution 50
Colour Reference Solution 60
Colour Reference Solution 70
Colour Reference Solution 100
Colour Reference Solution 150
Colour Reference Solution 200
Colour Reference Solution 250
Colour Reference Solution 300
Colour Reference Solution 350
Colour Reference Solution 400
Colour Reference Solution 450
Colour Reference Solution 500
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge