Seal roller, QuickSeal Hand Roller™
Lieferant: iST Scientific Ltd
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
IST-202-102HR
731-0461EA 73.1 EUR InStock
731-0461
Seal roller, QuickSeal Hand Roller™
Filmaufziehgeräte Verschlussfolienapplikator
This adhesive seal roller ensures even pressure is applied across the seal for a complete application to the plate.
The handle of the roller is made of a durable plastic, with a semi-hard padded rolling wheel.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge