Heat-Sealing-Folie, ClearASeal Peel™
Lieferant: iST Scientific Ltd
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
IST-101-115LR IST-101-078LR IST-101-078LS IST-101-078SR IST-101-078SS IST-101-078TR IST-101-078TS IST-101-115SR IST-101-115TR
731-0477EA 558 EUR InStock
731-0477 731-0475 731-0479 731-0476 731-0480 731-0481 731-0483 731-0478 731-0482
Heat-Sealing-Folie, ClearASeal Peel™
Versiegeln Mikrotiterplatten-Dichtungen
Heat sealing offers a 100% effective method for plate sealing for a complete seal integrity, as well as being quick and cost effective. The ClearASeal Peel is an optically clear laminate film forming a peelable seal to polypropylene, polyethylene, polystyrene, polycarbonate and cyclic olefin copolymer (COC) plates
  • Moderate solvent resistance, can be utilized for short term compound storage at room temperature
  • Available as sheets, for use with manual and semi-automated sealers, or in multiple roll formats compatible with specified automated heat sealers
  • Sheet seals are inter-leaved with paper sheets to help denote which side is the sealing side and aid removal of one sheet at a time from the pack
The optical clarity of this seal enables its use for sealing plates required for imaging use, including fluorescent detection methods such as qPCR and colorimetric assays.
The ClearASeal Peel forms a complete seal to a plate enabling both low temperature uses, including low temperature storage, and high temperature uses, such as PCR (when used with a pressurized heated lid).
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge