Kryoboxen für 0,2 ml PCR tubes
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F188370000
731-0703EA 149 EUR InStock
731-0703
Kryoboxen für 0,2 ml PCR tubes
Boxen Kryoboxen
These storage boxes are designed to fit in a standard 2" freezer racks. This 144-place PCR tube freezer box will hold individual and/or strip 0,2 ml PCR tubes.
  • Designed for ULT freezer storage at temperatures down to −86 °C
  • Durable and impervious to moisture
  • Clear polycarbonate lid and base and alpha-numeric index make it easy to identify samples within the box
  • Two boxes may be stacked to fit together in one space in a 2" freezer storage rack if using flat-capped PCR tubes
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge