PCR plates, 96-well, full skirt, Axygen®
Lieferant: Corning
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PCR-96-FS-B
731-4094EA 67.83 EUR InStock
731-4094
PCR plates, 96-well, full skirt, Axygen®
Mikrotiterplatten PCR-Platten
These Axygen® 96 well full skirt PCR microplates are designed to fit standard 96 well blocks. Ultra-thin and consistent wall thickness allows precise thermal transfer for optimum results. Automation friendly full skirt design can be sealed with 8/12 strip caps, Axymat or sealing film.
  • RNase-/DNase-free
  • Non pyrogenic, non sterile
  • Suitable for 0,1 ml thermal cycler blocks
  • Single notch corner
These plates are manufactured with 99,9% pure virgin PP.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge