Organic reference standard, Bisphenol A (3-chloro-2-hydroxypropyl) glycidyl ether, analytical standard, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
73124-100MG
73124-100MGEA 326 EUR InStock
73124-100MG
Organic reference standard, Bisphenol A (3-chloro-2-hydroxypropyl) glycidyl ether, analytical standard, Supelco®
Standards Organische Standards
Bisphenol A (3-chloro-2-hydroxypropyl) glycidyl ether is a chlorinated derivative, which is released during the thermal coating treatment of food cans, big storage vessels and food containers.
Bisphenol A (3-chloro-2-hydroxypropyl) glycidyl ether may be used as a reference standard for the determination of bisphenol A (3-chloro-2-hydroxypropyl) glycidyl ether in canned food and beverages using fast liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS).
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20733d2dfb4f9d751adff4c4c9eae1b6c4ef305f&vid=20733d2dfb4f9d751adff4c4c9eae1b6c4ef305f&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge