PCR microplates, 384-well Rigi-Plate, Axygen®
Lieferant: Corning
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PCR-384-RGD-BK
732-0952EA 203 EUR InStock
732-0952
PCR microplates, 384-well Rigi-Plate, Axygen®
Mikrotiterplatten PCR-Platten
Flat bottom, deep well microplates are designed to provide extra rigidity in thermal cycling applications.
Constructed of sturdy and durable polypropylene, plates are available in three color styles for application specific procedures. Choice of clear, black, or white plates allow for colorimetric, fluorescent, or luminescent applications. Certified RNase-, DNase-, and pyrogen-free. Well volume: 40 µl.
Bestellinformation: Clear plates are packaged 20 plates per pack, 5 packs per case. Black and white plates are packaged in sleeves of 20, with 5 sleeves per case.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge