Deep well and assay plates, Axygen®
Lieferant: Corning
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
P-2ML-SQ-C P-2ML-SQ-C-S P-384-240SQC P-384240SQCS P-5ML-48-C P-5ML-48-C-S P-96-450R-C P-96-450R-C-S P-96-450V-C P-96-450V-C-S P-DW-11-C-S P-DW-20CSIND P-DW-500-C P-DW-500-C-S P-384-120SQ-C P-DW-11-C P-DW-20-C P-DW-20-C-S P-DW-12-HC P-DW-12-HC-S
736-0344EA 131 EUR InStock
736-0344 736-0345 736-0332 738-1000 736-0348 736-0349 732-1370 732-1371 732-1372 732-1373 736-0339 738-0187 736-0335 736-0336 736-0357 736-0358 736-0359 737-0159 738-0185 738-0186
Deep well and assay plates, Axygen®
Mikrotiterplatten Deep-Well-Mikroplatten
Axygen® deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage.
  • 0,6 ml deep well plate
  • ANSI/SLAS 1-2004: Microplates - footprint dimensions
  • Ultra-flat surface ensures proper sealing with heat sealing film
Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to −80 °C.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge