VWR®, Deep Well Plates, Square Well
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
732-3323EA 247 EUR InStock
732-3323 732-3325 732-3795 732-3796 732-3797 732-3806 732-3810 732-3811 732-3866
VWR®, Deep Well Plates, Square Well
Mikrotiterplatten Deep-Well-Mikroplatten
These deep square well plates, made from virgin PP, are designed to make the interchanging of plates simpler in automated systems. They are also ideal for housing both cell and bacterial culture.
  • Chemical resistant
  • Free from DNase and RNase
  • V-bottom well shape maximises liquid removal and aids resuspension
  • U bottom suited for washing or stirring samples
  • No inner edges to interfere with magnetic bead collection
  • Raised well rims to improve heat sealing
  • Conical base aids sample concentration, reconstitution and centrifugation
Minimum order quantity of 40 cases for barcoded plates.
Achtung! Bitte beachten: Can be autoclaved once, but only if not used with robotic systems.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge