VWR®, Deep Well Plates, 96 Round Well
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
732-3322EA 266 EUR InStock
732-3322 732-3324 732-3328 732-3716 732-3718 732-3719 732-3802 732-3803
VWR®, Deep Well Plates, 96 Round Well
Mikrotiterplatten Deep-Well-Mikroplatten
Made of virgin PP to minimise extractables.
  • Raised rim to prevent cross contamination and enable a better heat-welded seal
  • Also available rimless to allow insertion of vials, or when adhesive seals are used
  • Free from DNase and RNase
  • Round bottom for optimal mixing and recovery
  • Small volumes with flat, U- or V-bottoms are also available
  • Can be stored at –80 °C
Suitable for use with automated sample handling systems.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge