Brilliant III SYBR® MM, Agilent Technologies
Lieferant: AGILENT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
600882 600883 600886 600887
732-3799EA 429 EUR InStock
732-3799 AGLS600883 AGLS600886 AGLS600887
Brilliant III SYBR® MM, Agilent Technologies
Nukleinsäure-Reagenzien qPCR/RT-qPCR Enzyme und Kits
Brilliant III Ultra-Fast SYBR® QPCR and QRT-PCR Master Mixes deliver superior specificity by utilizing a novel faster-activating hot start method to minimize the formation of primer-dimers and other off-target reactions.
  • Reliable and reproducible data
  • Novel fast Taq mutant for qPCR results in under 35 minutes
  • Enhanced rapid hot start capability saves time and reduces primer-dimer formation
  • Convenient pre-blended formulations compatible with any SYBR Green dye
These unique reagents are equally robust and reproducible across a variety of assays on fast cycling real-time platforms.
Achtung! Bitte beachten: For research use only. Not for use in diagnostic procedures.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge