Automated western device, eZwest™ Lite
Lieferant: Genscript
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
L00816
732-3879EA 5550 EUR InStock
732-3879
Automated western device, eZwest™ Lite
Blotting-Apparate
eZwest™ Lite is an automated western blotting device providing an easy and simple solution in your western blotting workflow. After proteins are transferred onto the membrane, load the membrane into our eZwest™ Lite device for automated blocking, incubation, and washing cycles. The user only needs to prepare the required reagents, run the customised program, and eZwest™ Lite will take care of the rest. Once the program is finished, the user can collect the results and immediately proceed to detection and analysis steps. eZwest™ Lite is easy to use and has minimal footprint.
  • Consistent Results: Fully automated operation ensuring consistency every time
  • Antibody Saving: Automatically recover primary antibody for re-use
  • Programmable: Easily customization program for protocol need
  • Completely Hands-off: Load your reagents and Go
eZwest™ produces similar or better results compared to results from the conventional method. Additionally, eZwest™ delivers consistent and reproducible results every time. The eZwest™ Lite device is compatible with the traditional reagents and protocols used for conventional western blotting. No proprietary reagents are required. Fully customisable steps and solutions make eZwest™ Lite a powerful tool fitting specific experiment needs for both routine and novel western blotting conditions.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge