Precast gels, Latitude™ HT Gels and Midigels
Lieferant: LONZA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
57220 57226 57246 57224 57230 57231 57234
733-1740EA 328 EUR InStock
733-1740 733-1741 733-1743 733-1596 733-1598 733-1599 733-1601
Precast gels, Latitude™ HT Gels and Midigels
Elektrophorese-Gele Fertiggele
Latitude™ DNA gels are designed for accurate DNA fragment separation for almost any gel electrophoresis application. These ready-to-run gels are available in a variety of sizes and well formats to fit most high-throughput needs.
  • Made with high quality, molecular biology grade SeaKem® or NuSieve® agarose
  • DNase- and RNase-free
  • Provide sharp bands with low background fluorescence
  • Ideal for checking restriction digests and PCR products
  • Fit in most large submerged electrophoresis systems and are compatible with multichannel pipettors
Both the DNA HT gels and the DNA Midigels are pre-stained with 0,5 µg/ml ethidium bromide.
Lieferumfang: Latitude™ HT gels are packaged 5 gels/box. Midigels are packaged 8 gels/box.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge