Magnetic separation devices, Axygen®
Lieferant: Corning
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
IMAG-12T IMAG-96P
733-2133EA 971.28 EUR InStock
733-2133 733-2134
Magnetic separation devices, Axygen®
Gestelle Trenngestelle, magnetisch
The magnetic separation devices (MSD) are essential in any paramagnetic bead-based purification process. These IMAG™ devices are designed for a manual process.
  • Single tube or 96-well format
  • Applications include nucleic acid purification, cell based assays and antobody and protein purifications
  • Strong magnets enable separation times of less than 30 seconds
  • Manual operation for either low or high throughput
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge