sparQ PureMag beads
Lieferant: QIAGEN BEVERLY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
95196-450 95196-060 95196-005
733-2628EA 4030 EUR InStock
733-2628 733-2627 733-2626
sparQ PureMag beads
Nukleinsäure-Reagenzien Next Generation Sequencing Reagenzien
sparQ PureMag beads is a fast and reliable nucleic acid purification system for reaction cleanup and size selection in Next Generation Sequencing (NGS) workflows. Based on the reversible nucleic acid-binding properties of magnetic beads; this product can be used to quickly remove primers, primer-dimers, unincorporated nucleotides, salts, adapters and adapter-dimers from NGS library prep reactions to improve downstream sequencing performance. sparQ PureMag beads allows excellent recovery of fragments greater than 100 bp without centrifugation or filtration. Consistent and reliable size selection can be achieved by simply adjusting the beads to sample ratio. This product is designed for both manual and automated processing, allowing seamless integration into existing workflows.
  • High recovery of DNA fragments greater than 100 bp
  • Efficient removal of unwanted components from adapter ligation and PCR reactions
  • Consistent single- or double-sided size selection
  • Seamless integration into existing NGS workflows with little or no protocol change
  • Easy to use and compatible with manual processing or automated liquid handling robots
  • Cost effective alternative to AMPure® XP with equivalent performance
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge