eQo One-Step ToughMix®
Lieferant: QIAGEN BEVERLY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
95301-100 95301-01K
733-2941EA 319 EUR InStock
733-2941 733-2940
eQo One-Step ToughMix®
Nukleinsäure-Reagenzien qPCR/RT-qPCR Enzyme und Kits
No compromises. Unparalleled performance meets sustainability.
  • Unrivaled sensitivity - detection down to two copies of RNA
  • Sustainable and earth friendly: Lyophilised eQo bead format eliminates dry ice, CO₂ consumptions and reduces shipping costs
  • Novel reverse transcriptase: Convenient, ambient reaction setup, overcomes rigid RNA secondary structure
  • High stability: Up to one year at room temperature, up to one year rehydrated at −20 °C
  • Tough Tested: Broad and significantly enhanced tolerance to PCR inhibitors
eQo 1-step ToughMix is a lyophilised reagent system for reverse transcription quantitative PCR (RT-qPCR) of RNA templates using hybridisation probe detection chemistries such as TaqMan® 5’-hydrolysis probes. It is supplied with a proprietary rehydration buffer that when combined with the lyophilised 'eQo beads' produces a stabilised 4X concentrated master mix.
The kit includes thermolabile UDG for amplicon carryover elimination, an enhanced warm-start reverse transcriptase (RT) and all other required components for 1-step RT-qPCR except RNA template and primer/probe(s). The reaction chemistry has been optimised for inhibitor tolerance and delivers exceptional performance in either single-plex or highly demanding multiplex 1 step RT-qPCR formats.
Cycler compatibility: BioRad CFX, Quantabio Q, Roche LightCycler 480
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge