95301-100
95301-01K
733-2941EA
319
EUR
InStock
733-2941
733-2940
eQo One-Step ToughMix®
Nukleinsäure-Reagenzien
qPCR/RT-qPCR Enzyme und Kits
No compromises. Unparalleled performance meets sustainability.
- Unrivaled sensitivity - detection down to two copies of RNA
- Sustainable and earth friendly: Lyophilised eQo bead format eliminates dry ice, CO₂ consumptions and reduces shipping costs
- Novel reverse transcriptase: Convenient, ambient reaction setup, overcomes rigid RNA secondary structure
- High stability: Up to one year at room temperature, up to one year rehydrated at −20 °C
- Tough Tested: Broad and significantly enhanced tolerance to PCR inhibitors
eQo 1-step ToughMix is a lyophilised reagent system for reverse transcription quantitative PCR (RT-qPCR) of RNA templates using hybridisation probe detection chemistries such as TaqMan® 5’-hydrolysis probes. It is supplied with a proprietary rehydration buffer that when combined with the lyophilised 'eQo beads' produces a stabilised 4X concentrated master mix.
The kit includes thermolabile UDG for amplicon carryover elimination, an enhanced warm-start reverse transcriptase (RT) and all other required components for 1-step RT-qPCR except RNA template and primer/probe(s). The reaction chemistry has been optimised for inhibitor tolerance and delivers exceptional performance in either single-plex or highly demanding multiplex 1 step RT-qPCR formats.
Cycler compatibility: BioRad CFX, Quantabio Q, Roche LightCycler 480