Fibronektin-Zellkultureinsätze, Corning® BioCoat™ FluoroBlok™
Lieferant: Corning
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
354597
734-1053EA 625 EUR InStock
734-1053
Fibronektin-Zellkultureinsätze, Corning® BioCoat™ FluoroBlok™
Mikrotiterplatten Einsetz-Platten und Einsätze
Corning® BioCoat™ FluoroBlok™ fibronectin-coated cell culture inserts packaged ready to use in 24-well Falcon® companion plates.
  • Convenient, individual inserts packaged in a one-piece, automation friendly 24-multiwell format
  • Combined benefits of Falcon® inserts coated with HFN for enhanced cell attachment and growth
  • Individual inserts contain a 3,0 µm pore size clear PET membrane
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge