HTS Caco-2 assay system, PET membrane, BioCoat™, Corning
Lieferant: Corning
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
354802
734-1078EA 1790.09 EUR InStock
734-1078
HTS Caco-2 assay system, PET membrane, BioCoat™, Corning
Mikrotiterplatten Zellkultur-Platten
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.
Corning BioCoat HTS Caco-2 assay system, PET membrane contains specially formulated serum-free medium, culture supplements, sodium butyrate, and the Corning BioCoat fibrillar collagen 24-well insert system.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge