Dialyseeinheiten, Tube-A-Lyzer®, Spectra/Por®
Lieferant: Spectrum Laboratories
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
137009 137042 137046 137050
734-1451EA 302 EUR InStock
734-1451 734-1453 734-1455 734-1458
Dialyseeinheiten, Tube-A-Lyzer®, Spectra/Por®
Schläuche Dialyseschläuche
Disposable and ready-to-use dynamic dialysis device.
  • Faster dialysis (4 to 12 hours)
  • Easy sample loading, testing and recovery
  • Prevents membrane polarisation and fouling
  • No heavy metals and sulphide contaminant
Device incorporates a semi-permeable membrane tubing that separates the sample chamber from the surrounding flow-through buffer chamber. The housing is designed to be a flow-through dialysate chamber that can be connected to a buffer source and a waste collection vessel or discharging directly to drain.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge