Deckel für 96-Well-PCR-Platte, Corning® Thermowell®
Lieferant: Corning
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
6520
734-1754EA 136.69 EUR InStock
734-1754
Deckel für 96-Well-PCR-Platte, Corning® Thermowell®
Deckel Deckel für Mikrotiterplatten
Polycarbonate lids are thermally stable up to 135 °C and can be used on Corning® Thermowell™ 96 well polycarbonate plates (fits Cat. No. 6511) with oil overlays.
Lids are intended to be covers only and do not prevent evaporation.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge