Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Cellulose Soxhlet extraction thimbles, Whatman™

Lieferant: GE Healthcare - Whatman

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

2800-105 2800-185 2800-199 2800-226 2800-228 2800-250 2800-258 2800-266 2800-280 2800-282 2800-288 2800-300 2800-308 2800-330 2800-331 2800-338 2800-339 2800-373 2800-412 2800-432 2800-608 2810-228 2810-258 2810-338 2810-432 2810-339 2800-259 2800-260 2810-166 2810-266 2810-902
734-2500EA 111 EUR InStock
734-2500 734-2501 734-2502 734-2578 734-2503 734-2504 734-2505 734-2506 734-2507 734-2508 734-2509 734-2510 734-2511 734-2512 734-2513 734-2514 734-2515 734-2516 734-2517 734-2518 734-2519 515-0167 515-0168 515-0169 515-8943 537-0036 734-2579 734-2580 537-0034 537-0035 WHAT2810-902
Cellulose Soxhlet extraction thimbles, Whatman™
Extraktoren Extraktionshülsen
Cytiva's Whatman cellulose thimbles are manufactured from high-quality alpha cellulose cotton linter for high-performance use in automated Soxhlet extractor systems. Used in air and waste gas analysis.

  • Used in a wide variety of solid-liquid extraction studies, including lipid and pesticide analyses in food and soil samples
  • Suitable for temperatures up to 120 °C (248 °F)
  • Excellent mechanical strength and retention
  • Wall thickness of standard thimbles is 1 to 1,5 mm to support application need
  • Option for double thickness for added rigidity

Standard extraction thimbles have a wall thickness of 1 to 1,5 mm. Double-thickness thimbles have a wall thickness of approximately 2 mm for applications that require higher retention, increased wet or dry strength, or increased rigidity. Measurements can be matched to specific Soxhlet extractor systems.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge