Racks, magnetic separation, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F199000000 F199000001 F199000002 F199000003 F199000004
734-2889EA 1120 EUR InStock
734-2889 734-2890 734-2891 734-2892 734-2893
Racks, magnetic separation, SP Bel-Art
Gestelle Trenngestelle, magnetisch
Acrylic, clear.
  • Independent sleeves make liquid addition and removal easier with less accidental fluid loss
  • Powerful magnets quickly secure beads
  • Spring plunger on tube sleeves allows beads to be sequestered at various heights along the tube wall
  • Simply turn tube for efficient bead mixing
These magnetic separation racks has sequestered magnetic beads, so that rinse solutions and affinity ligands such as antibodies, streptavidin, or proprietary reagents for coupling nucleic acids can be added or decanted in assays. The 96-well PCR plate model has 24 magnets that snugly hold the plate while attracting beads to the wall of the tubes, whereas flat bottom microplate model has magnets covering entire footprint to accept all standard micro-plates with any number of wells. (i.e.,) 6, 12, 24, 48, 96.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge