Avantor® Rigid Outer Support Containers
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
734-3421EA 1580 EUR InStock
734-3421 734-3422 734-3423 734-3424 734-3425 734-3426 734-3427 734-3428 734-3429 734-3430 734-3431 734-3432
Avantor® Rigid Outer Support Containers
Beutel Aufbewahrungsbeutel
3D cubic bags can fit in existing support containers. These support containers are available in either High Density Polyethylene (HDPE) and Polypropelene (PP), or stainless steel material.
  • Standard 3D cubic bags are available in 100, 200, and 1000 L volumes with 3 top ports
  • Standard 3D cubic bags are available with or without a bottom port
  • Cleanroom and shipping variants are available in either material format
  • Plastic containers can collapse for easy storage
  • Stainless steel containers are ISTA certified for shipping
Avantor® 3D cubic bags are made from a proprietary film which features a polyethelene inner and outer layer with gas and moisture barrier layer between. The film has high capacity and flexibility, and is resistant to a wide range of chemicals.
Zertifizierungen: Stainless steel containers meet ISTA 3,1B certification.
Order Now
PRODUKT
TECHNISCHE DATEN
EGÄNZENDE PRODUKTE
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge