MultiScreen Filter Plates with Hydrophilic Durapore® PVDF Membrane
Lieferant: Merck
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
MAHVN4550 MADVN6550 MAGVS2210 MAHVN4510 MABVN1250 MABVN0B50
736-0186EA 1600 EUR InStock
736-0186 736-0177 736-0181 736-0185 736-0175 MILFMABVN0B50
MultiScreen Filter Plates with Hydrophilic Durapore® PVDF Membrane
Mikrotiterplatten Mikrofiltrationsplatten
MultiScreen HTS filtration plates are used to filter samples and perform entire procedures, from cell growth to scintillation counting within the same plate.
  • Hydrophilic Durapore® polyvinylidene fluoride (PVDF) membrane
The plates come in many membrane types, pore sizes, and plate materials. The automation-compatible design allows for easy manipulation with a variety of robotics systems and makes barcoding possible on all four sides. Filtration is accomplished either by vacuum using the MultiScreen HTS Vacuum Manifold, or by centrifugation.
MultiScreenHTS plates are specifically developed for high-throughput use with automated work stations. Rigid sidewalls improve handling and ample surfaces are provided for bar code labels. Wells are individually sealed to prevent incubation crosstalk and the underdrain is removable for access to the filters.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge