Axygen® Robotic Tips for Corbett X-tractor
Lieferant: Corning
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
CTF-199-R-S
736-0522EA 591 EUR InStock
736-0522
Axygen® Robotic Tips for Corbett X-tractor
Pipettenspitzen
Tips in 96-well format designed for Corbett X-tractor Gene 8 channel head.
  • Clear polypropylene
  • With aerosol-resistant filters
  • Optimised fine-point for accurate dispensing
  • Certified RNase, DNase and endotoxin-free
These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge