Microplates, 1536-Well
Lieferant: GREINER BIO ONE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
782061_60 782074_60 782077_60 783096_60 783076_60 782075_60 782076_60 782095_60 782096_60 782101_60
392-0138EA 2450 EUR InStock
392-0138 738-0067 738-0070 738-0089 738-0084 738-0068 738-0069 738-0077 738-0078 738-0080
Microplates, 1536-Well
Mikrotiterplatten HTS-Mikrotiterplatten
PS, without lid. Ideal for high throughput screening applications, these plates are available as clear bottom variants (in clear PS) and black or white in LUMITRAC™ or FLUOTRAC™ quality.
  • HiBase versions designed for top reading instruments, µClear® LoBase plates for bottom reading instruments
  • Solid bottom plates available with high binding (MICROLON™ 600, LUMITRAC™ 600 or FLUOTRAC™ 600) or medium binding (LUMITRAC™ 200 or FLUOTRAC™ 200) surfaces
  • Rounded square well design combines the advantages of the square well (flexible working volume, 3 to 10 µl) with the advantages of a round well (reduced wicking and bubbling)
  • Free of DNase, RNase and human DNA, and are non-pyrogenic
Working volume: 3 - 10 µl
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge