Microplates, 96-well, non binding
Lieferant: GREINER BIO ONE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
650901_40 655901_40 655900_40 655906_40 655903_40 655904_40 651901_40
738-0165EA 270 EUR InStock
738-0165 738-0170 738-0169 738-0173 738-0171 738-0172 738-0166
Microplates, 96-well, non binding
Mikrotiterplatten HTS-Mikrotiterplatten
Characterised by low protein, DNA, RNA, and peptide binding properties, the non binding microplate surface significantly increases assay sensitivity by reducing background and improving signal-to-noise ratio.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge