Microplates, 384-well, non binding
Lieferant: GREINER BIO ONE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
781900_40 784900_40 781906_40 781904_40 781903_40 784904_40 781901_40
738-0175EA 377 EUR InStock
738-0175 738-0179 738-0178 738-0177 738-0176 GREI784904_40 GREI781901_40
Microplates, 384-well, non binding
Mikrotiterplatten HTS-Mikrotiterplatten
Characterised by low protein, DNA, RNA, and peptide binding properties, the non binding microplate surface significantly increases assay sensitivity by reducing background and improving signal-to-noise ratio.
All plates are free from RNase, DNase, and pyrogens, and are also free of DNA polymerase inhibitors.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge