Organic reference standard, Padimate O, analytical standard, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
74309-1ML
74309-1MLEA 101 EUR InStock
74309-1ML
Organic reference standard, Padimate O, analytical standard, Supelco®
Standards Organische Standards
Padimate O is a tertiary amine derivative of p-aminobenzoic acid (PABA), which is commonly used as a sunscreen agent in cosmetics and other sunscreen formulations due to its ability to absorb ultraviolet radiations.
Padimate O may be used as a reference standard in the determination of padimate O in cosmetics using high performance liquid chromatography (HPLC).
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff82f7d9a36fa3b985395f274f6d04abefaa7dd9&vid=ff82f7d9a36fa3b985395f274f6d04abefaa7dd9&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge