Micro particles based on polystyrene, analytical standard, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
74491-10ML-F 74491-5ML-F 74964-10ML-F 74964-5ML-F 79807-5ML-F 84135-10ML-F 84135-5ML-F
74491-10ML-FEA 531 EUR InStock
74491-10ML-F 74491-5ML-F 74964-10ML-F 74964-5ML-F 79807-5ML-F 84135-10ML-F 84135-5ML-F
Micro particles based on polystyrene, analytical standard, Supelco®
Standards Partikelgrößenanalyse-Standards
For every lot exact values of particle size and standard deviation are determined with an accuracy of 0,01 μm using a Coulter Multisizer.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge