Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Flashpoint testers, Setaflash 3

Lieferant: STANHOPE SETA

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

31000-0 33000-0 33250-2 33200-2U
751-0013EA 0 EUR InStock
751-0013 751-0065 751-0067 751-0066
Flashpoint testers, Setaflash 3
Flammpunktprüfer
Four versions of the closed cup flashpoint testers are available. All functions are accessed by a two-button keypad and the SMART control feature (Seta Multifunctional And Rotational Test). The digital display shows test parameters, instrument status and the test results. Audible prompts are sounded when an action is required by the user. The automatic flash detector reduces the chance of misinterpretation of the results and enhances repeatability. Instruments have a integral gas tank which is filled using a standard butane (lighter) refill cartridge and are suitable for mobile use when operated with the optional 12 volt power supply adapter ( please ask for details).

  • Flash point tests in less than two minutes
  • Small sample sizes of 2 or 4 ml
  • Audible and on screen prompts for ease of use

Closed cup tester: As above with the addition of a temperature ramp mode that allow the rapid determination of the flash point of a sample in accordance with ASTM D7236 and IP 534.

Active cool closed cup tester: Active Cool function to rapidly and efficiently carry out Flash/No flash tests and extend the temperature down to 10 °C - Cooling and heating with Peltier cells.

Active cool corrosion resisting closed cup tester: Flash/No Flash testing (10 to 130 °C) rapid temperature normalisation between tests. Corrosion resisting insert in the sample cup.

33250-2 Setaflash 3 Active Cool (corrosion resisting) closed cup tester: Flash/No Flash testing (10 to 130 °C) rapid temperature normalisation between tests. Corrosion resisting insert in the sample cup.

Zertifizierungen: Basic models: ASTM D3278, BS EN456, ISO 3680, UN class 3 non viscous flammable liquids, ASTM E502, ISO 3679, BS 3900 Part A13, EN 456, ASTM D3828, IP 303-1/2, CHIPS regulations, classification of dangerous goods for carriage: Viscous and non viscous liquids, EPA 1020 A/B, IP 523, 524.
Models with ramp mode. As above plus ASTM D7236 and IP 534.

Lieferumfang: Supplied with a syringe, mains lead and instruction manual.

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge